missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Catenin, p Ser33, p Ser37 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24456
This item is not returnable.
View return policy
Description
beta-Catenin Polyclonal specifically detects beta-Catenin in Human samples. It is validated for Western Blot.
Specifications
| beta-Catenin (p Ser33, p Ser37) | |
| Polyclonal | |
| Unconjugated | |
| 0.1 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, DKFZp686D02253, FLJ25606, FLJ37923 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen for this SMO antibody is 100% homologous in human, monkey and mouse. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.2 mg/mL | |
| Western Blot 0.25-1.0 ug/ml | |
| P35222 | |
| CTNNB1 | |
| A portion of human Beta-catenin protein containing phosphorylated serine residues at position 33 and 37 was used as the immunogen for this phospho antibody. | |
| 0.02 mg | |
| Cancer, Cellular Markers, Extracellular Matrix, Neuroscience, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
| 1499 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction