Learn More
beta Catenin Monoclonal Antibody (15B8), eFluor™ 660, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£81.00 - £205.00
Specifications
Antigen | beta Catenin |
---|---|
Clone | 15B8 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15589916
|
Affymetrix eBioscience
50-2567-42 |
100 Tests |
£205.00
100 tests |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15579916
|
Affymetrix eBioscience
50-2567-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The 15B8 monoclonal antibody reacts with human and mouse beta-catenin, one member of a family of catenins, which are intracellular proteins that interact with cadherins to mediate cellular adhesion. More specifically, beta-catenin binds to the cytoplasmic tail of E-cadherin. In addition, this molecule is a component of the canonical Wnt signaling pathway. In the absence of Wnt binding its receptor, beta-catenin is phosphorylated and resides in the cytoplasm where it is eventually targeted for degradation by ubiquitination. Upon Wnt binding, beta-catenin becomes dephosphorylated, translocates to the nucleus, and modulates gene expression in partnership with the transcription factors T cell factor (TCF) and lymphocyte enhancer binding factor (LEF). Expression of beta-catenin is found in a wide variety of non-immune and immune tissues, including thymocytes and T and B lymphocytes. The Wnt and beta-catenin signaling pathway has been demonstrated to play a crucial role in the development of T, B, and hematopoietic stem cells. Applications Reported: This 15B8 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This 15B8 antibody has been tested by intracellular staining and flow cytometric analysis of the Jurkat cell line using the Foxp3/Transcription Factor Staining Buffer Set (cat. 00-5523). This can be used at less than or equal to 0.25 μg per test.
beta Catenin Monoclonal antibody specifically detects beta Catenin in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
beta Catenin | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P35222, Q02248 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
15B8 | |
Flow Cytometry | |
eFluor 660 | |
Mouse | |
Human, Mouse | |
12387, 1499 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
CTNNB1 |
For Research Use Only.