missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
beta-Catenin Monoclonal antibody specifically detects beta-Catenin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | beta-Catenin |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | GT2169 |
| Conjugate | Unconjugated |
| Formulation | PBS |
| Gene Alias | beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, DKFZp686D02253, FLJ25606, FLJ37923 |
| Host Species | Mouse |
| Immunogen | Recombinant protein encompassing a sequence within the N-terminus region of human beta-Catenin. The exact sequence is proprietary. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?