missing translation for 'onlineSavingsMsg'
Learn More

beta-Catenin Antibody (CTNNB1/8280R), Novus Biologicals™

Rabbit Monoclonal Antibody

£205.00 - £377.00

Specifications

Antigen beta-Catenin
Clone CTNNB1/8280R
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin : 1-2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18655064
View Documents
Novus Biologicals
NBP3-23721-100ug
100 μg
£377.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18671104
View Documents Promotion Details
Novus Biologicals
NBP3-23721-20ug
20 μg
£205.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

beta-Catenin Monoclonal antibody specifically detects beta-Catenin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

beta-Catenin
0.2 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Immune System Diseases, Mucosal Immunology, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway
10 mM PBS with 0.05% BSA
1499
IgG κ
Protein A purified
CTNNB1/8280R
Immunohistochemistry-Paraffin : 1-2 μg/mL
Monoclonal
Purified
RUO
Human
beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, DKFZp686D02253, FLJ25606, FLJ37923
Recombinant fragment (around aa1-200) of human beta-Catenin protein (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.