missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
beta-Catenin Antibody (CTNNB1/1509), Novus Biologicals™
Mouse Monoclonal Antibody
£221.00 - £400.00
Tekniske data
| Antigen | beta-Catenin |
|---|---|
| Clone | CTNNB1/1509 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Quantity | Pris | Antal & tilgængelighed | |||||
|
18231230
|
Novus Biologicals
NBP2-53366-20UG |
20 μg |
£221.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18246470
|
Novus Biologicals
NBP2-53366-100UG |
100 μg |
£400.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivelse
beta-Catenin Monoclonal specifically detects beta-Catenin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Tekniske data
| beta-Catenin | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Cellular Markers, Extracellular Matrix, Mucosal Immunology, Neuroscience, Phospho Specific, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| 1499 | |
| Recombinant human full-length beta-Catenin protein fragment | |
| Primary | |
| Store at 4C. |
| CTNNB1/1509 | |
| Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, DKFZp686D02253, FLJ25606, FLJ37923 | |
| CTNNB1 | |
| IgG1 κ | |
| Protein A or G purified | |
| 92 kDa |
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel