missing translation for 'onlineSavingsMsg'
Learn More

beta-Catenin Antibody (CTNNB1/1508), DyLight 550, Novus Biologicals™

Product Code. 18412674 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18412674 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18412674 Supplier Novus Biologicals Supplier No. NBP254541R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

beta-Catenin Monoclonal antibody specifically detects beta-Catenin in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta-Catenin
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone CTNNB1/1508
Conjugate DyLight 550
Formulation 50 mM Sodium Borate
Gene Alias beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, DKFZp686D02253, FLJ25606, FLJ37923
Host Species Mouse
Immunogen Recombinant human beta-Catenin protein fragment (Uniprot: P35222)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cellular Markers, Extracellular Matrix, Mucosal Immunology, Neuroscience, Stem Cell Signaling Pathway, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 1499
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.