missing translation for 'onlineSavingsMsg'
Learn More

beta-Catenin Antibody (15B8), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18425574 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425574 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18425574 Supplier Novus Biologicals Supplier No. NBP254544MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

beta-Catenin Monoclonal antibody specifically detects beta-Catenin in Human, Mouse, Rat, Bovine, Canine, Chicken samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Tekniske data

Antigen beta-Catenin
Applications Flow Cytometry
Classification Monoclonal
Clone 15B8
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, DKFZp686D02253, FLJ25606, FLJ37923
Host Species Mouse
Immunogen Recombinant chicken beta-Catenin (Uniprot: P35222)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Immune System Diseases, Mucosal Immunology, Neuroscience, Phospho Specific, Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 1499
Target Species Human, Mouse, Rat, Bovine, Canine, Chicken
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.