missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Casein Antibody (F20.14) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2720JF635
This item is not returnable.
View return policy
Description
beta Casein Monoclonal antibody specifically detects beta Casein in Human samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| beta Casein | |
| Monoclonal | |
| Janelia Fluor 635 | |
| beta-casein, CASB, casein beta | |
| Purified casein and a-lactalbumin. | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 1447 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot | |
| F20.14 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur