missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta B1 Crystallin Antibody (3D9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00001414-M03
This item is not returnable.
View return policy
Description
beta B1 Crystallin Monoclonal antibody specifically detects beta B1 Crystallin in Human samples. It is validated for Western Blot, ELISA
Specifications
| beta B1 Crystallin | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| Beta-B1 crystallin, beta-crystallin B1, CATCN3, crystallin, beta B1, eye lens structural protein | |
| CRYBB1 (NP_001878, 37 a.a. ~ 137 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. TTLAPTTVPITSAKAAELPPGNYRLVVFELENFQGRRAEFSGECSNLADRGFDRVRSIIVSAGPWVAFEQSNFRGEMFILEKGEYPRWNTWSSSYRSDRLM | |
| 0.1 mg | |
| Vision | |
| 1414 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2b κ |
| Western Blot, ELISA | |
| 3D9 | |
| Western Blot 1:500, ELISA | |
| NP_001878 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction