Learn More
beta-Arrestin 2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA1732
Code : LE
Additional Details : Weight : 0.01000kg
Description
PA1-732 detects beta-Arrestin 2 in rat, human and bovine samples. PA1-732 has been successfully used in Western blot procedures. By Western blot, this antibody detects ∽49 kDa and ∽47 kDa proteins representing pan Arrestin in rat and bovine retina as well as rat brain samples. The PA1-732 immunogen is a synthetic peptide corresponding to residues C D(384) D I V F E D F A R L R L K(397) of human beta-arrestin 2. This peptide (Cat. # PEP-281) is available for use in neutralization and control experiments.
Beta-arrestin-2, or ARRB2, is an adaptor protein belonging to the arrestin family with an N- arrestin domain and a C- arrestin domain. ARRB2 is a regulatory protein involved in heterotrimeric G protein-coupled receptor desensitization and is known to regulate beta2-adrenergic receptor (beta2AR) function by enhancing beta2AR mediated nuclear translocation of ERK. ARRB2 binds to phosphorylated beta2ARs, thereby causing a significant impairment of their capacity to activate G(S) proteins. Along with AIP4, ARRB2 acts as an endosomal sorting molecule that mediates CXCR4 entry into a degradative pathway. It may also be involved in hormone-specific desensitization of TSH receptors. ARRB2 is a Ca2+ binding protein of the retinal outer segments and binds to P-rhodopsin and is also involved in synaptic transmission in photoreceptor cells. It is widely expressed in most of the tissues, especially neuronal tissues and spleen.Specifications
beta-Arrestin 2 | |
Polyclonal | |
Unconjugated | |
ARRB2 | |
AI326910; ARB2; ARR2; ARRB2; arrestin 3; arrestin beta 2; arrestin beta-2; arrestin, beta 2; arrestin-3; AW122872; BARR2; BARRES; beta arr2; beta-arrestin 2 long isoform; beta-arrestin 2 short isoform; beta-arrestin-2; Non-visual arrestin-3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
216869, 25388, 281638, 409 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
P29067, P32120, P32121, Q91YI4 | |
ARRB2 | |
Synthetic peptide corresponding to residues C D(384) D I V F E D F A R L R L K(397) of human beta-arrestin 2. | |
100 μg | |
Primary | |
Bovine, Human, Mouse, Rat | |
Antibody | |
IgG |