Learn More
beta-Arrestin 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-587
This item is not returnable.
View return policy
Description
beta-Arrestin 2 Polyclonal specifically detects beta-Arrestin 2 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Western Blot, Immunoprecipitation | |
| Western Blot 1:2000, Immunoprecipitation 1:10 - 1:500 | |
| ARB2, ARR2, arrestin 3, Arrestin beta-2, arrestin, beta 2, BARR2, beta-arrestin-2, DKFZp686L0365 |