missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ beta-Arrestin 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29035
This item is not returnable.
View return policy
Description
beta-Arrestin 2 Polyclonal antibody specifically detects beta-Arrestin 2 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Western Blot
Specifications
| beta-Arrestin 2 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 409 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| ARB2, ARR2, arrestin 3, Arrestin beta-2, arrestin, beta 2, BARR2, beta-arrestin-2, DKFZp686L0365 | |
| Between 355 and 405 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction