missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Arrestin 2 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24569AF532
This item is not returnable.
View return policy
Description
beta-Arrestin 2 Polyclonal antibody specifically detects beta-Arrestin 2 in Human, Mouse, Rat, Bovine, Canine, Chicken, Equine, Primate, Xenopus, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| beta-Arrestin 2 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| ARB2, ARR2, arrestin 3, Arrestin beta-2, arrestin, beta 2, BARR2, beta-arrestin-2, DKFZp686L0365 | |
| A peptide sequence corresponding to an area between amino acids 1 and 50 of human ARRB2 was used as the immunogen for the antibody, Gen Bank no. gb:ABG47460.1. | |
| 0.1 mL | |
| Immunology, Virology Bacteria and Parasites | |
| 409 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Bovine, Canine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction