missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ beta Arrestin 1 Recombinant Rabbit Monoclonal Antibody (F7C2)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA537831
This item is not returnable.
View return policy
Description
Positive Samples: K-562, Mouse lung, Mouse spleen, Mouse kidney Immunogen sequence: MGDKGTRVFK KASPNGKLTV YLGKRDFVDH IDLVDPVDGV VLVDPEYLKE RRVYVTLTCA FRYGREDLDV LGLTFRKDLF VANVQSFPPA PEDKKPLTRL.
Beta Arrestin 1 is a member of a family of proteins widely expressed but especially abundant in the central nervous system. Serving as an adaptor or scaffold molecule, beta Arrestin 1 is essential for mitogenic signaling and mediates agonist-dependent desensitization and internalization of Gprotein-coupled receptors (GPCRs, e.g., beta 2-adrenergic receptor). After binding to their ligand and interacting with heterotrimeric G proteins, GPCRs are phosphorylated by G-protein receptor kinases (GRKs) on serine residues. Beta Arrestin 1 in the cytosol is phosphorylated by ERK1 and 2 on serine412 in a negative feedback mechanism and binds to the phosphorylated receptors at the plasma membrane. Serine 412 is then dephosphorylated and the GPCRs are internalized, leading to activation of the Ras, Raf, ERK1 and 2 signaling pathway.
Specifications
| beta Arrestin 1 | |
| Recombinant Monoclonal | |
| 1.33 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P49407, Q8BWG8 | |
| ARRB1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human beta-arrestin1 (P49407). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| F7C2 | |
| Unconjugated | |
| ARRB1 | |
| 1200006I17Rik; ARB1; ARR1; ARRB1; ARRB1 protein; arrdc1; arrestin; arrestin 2; arrestin beta 1; arrestin beta-1; arrestin, beta 1; arrestin-2; AW208571; BARRES; beta-arrestin; beta-arrestin1; beta-arrestin-1; G430100A01Rik; Non-visual arrestin-2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 109689, 408 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction