missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Arrestin 1 Rabbit anti-Human, Mouse, Clone: F7C2, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £367.00
Specifications
| Antigen | beta-Arrestin 1 |
|---|---|
| Clone | F7C2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18376496
|
Novus Biologicals
NBP3-15335-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18324184
|
Novus Biologicals
NBP3-15335-100UL |
100 μg |
£367.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
beta-Arrestin 1 Monoclonal antibody specifically detects beta-Arrestin 1 in Human, Mouse samples. It is validated for Western BlotSpecifications
| beta-Arrestin 1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction, Virology Bacteria and Parasites | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 408 | |
| IgG | |
| Affinity purified |
| F7C2 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| ARB1, ARR1, arrestin 2, Arrestin beta-1, arrestin, beta 1, beta-arrestin-1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human beta-Arrestin 1 (P49407). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title