Learn More
Beta Arrestin 1 Mouse anti-Human, Unconjugated, Clone: 1H8E4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67580-1-IG-20UL
Description
The Beta Arrestin 1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Beta Arrestin 1. This antibody recognizes human, mouse, rat antigen. The Beta Arrestin 1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| Beta Arrestin 1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, ELISA | |
| 1H8E4 | |
| Unconjugated | |
| 20 μL | |
| 408 | |
| IgG2a |