missing translation for 'onlineSavingsMsg'
Learn More

beta Amyloid Antibody (MOAB-2), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18414884
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414884 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414884 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP213075MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

beta Amyloid Monoclonal antibody specifically detects beta Amyloid in Human, Mouse, Rat, Bacteria, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot, Dot Blot, Immunohistochemistry Free-Floating
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta Amyloid
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone MOAB-2
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Dot Blot, Immunohistochemistry Free-Floating
Formulation 50mM Sodium Borate
Gene Alias Abeta, Amyloid beta, beta-amyloid peptide
Host Species Mouse
Immunogen This beta Amyloid antibody was developed against recombinant human beta Amyloid 42.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cardiovascular Biology, Cell Cycle and Replication, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 351
Target Species Human, Mouse, Rat, Bacteria, Monkey
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.