missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
beta Amyloid Monoclonal antibody specifically detects beta Amyloid in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | beta Amyloid |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | MM26R |
| Conjugate | DyLight 650 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Abeta, Amyloid beta, beta-amyloid peptide |
| Host Species | Mouse |
| Immunogen | Recombinant protein to human Amyloid β 1-42 peptide |
| Purification Method | Protein A or G purified from cell culture supernatant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?