missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Amyloid Antibody (donanemab), PE, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-27993PE
This item is not returnable.
View return policy
Description
beta Amyloid Monoclonal antibody specifically detects beta Amyloid in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| beta Amyloid | |
| Monoclonal | |
| PE | |
| Abeta, Amyloid beta, beta-amyloid peptide | |
| Amyloid Beta | |
| 0.1 mL | |
| Alzheimers Research, Apoptosis, Cardiovascular Biology, Cell Cycle and Replication, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific, Signal Transduction | |
| 351 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| donanemab | |
| PBS | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu