missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ PPP1R18 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29395
This item is not returnable.
View return policy
Description
PPP1R18 Polyclonal antibody specifically detects PPP1R18 in Human samples. It is validated for Immunoprecipitation
Specifications
| PPP1R18 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 170954 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation | |
| Unconjugated | |
| HKMT1098, KIAA1949, phostensin, PPP1R18, Protein Phosphatase 1 F-Actin Cytoskeleton Targeting Subunit, Protein Phosphatase 1 F-Actin Cytoskeleton-Targeting Subunit, Protein Phosphatase 1 Regulatory Subunit 18, Protein Phosphatase 1, Regulatory Subunit 18 | |
| Between 563 and 613 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto