missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ beta Amyloid Antibody (CNTO 2125), Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28180-1mg
This item is not returnable.
View return policy
Description
beta Amyloid Monoclonal antibody specifically detects beta Amyloid in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| beta Amyloid | |
| Monoclonal | |
| Unconjugated | |
| P05067 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| CNTO 2125 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| Abeta, Amyloid beta, beta-amyloid peptide | |
| Amyloid Beta | |
| 1 mg | |
| Alzheimers Research, Apoptosis, Cardiovascular Biology, Cell Cycle and Replication, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific, Signal Transduction | |
| 351 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction