missing translation for 'onlineSavingsMsg'
Learn More

beta Amyloid Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18408577 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408577 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408577 Supplier Novus Biologicals Supplier No. NBP225093AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

beta Amyloid Polyclonal antibody specifically detects beta Amyloid in Human, Mouse, Rat, Bacteria samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta Amyloid
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias Abeta, Amyloid beta, beta-amyloid peptide
Host Species Rabbit
Immunogen This beta Amyloid antibody was raised against a 10 amino acid peptide on the amino terminus of the 4KDa beta Amyloid peptide generated by beta- and gamma-secretases. The immunogen is located within amino acids 650 - 700 of human amyloid A4 protein precursor (APP). Amino Acid Squence: DAEFRHDSGYE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cardiovascular Biology, Cell Cycle and Replication, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 351
Target Species Human, Mouse, Rat, Bacteria
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.