missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ beta Amyloid 42 Peptide
Description
Amino Acid Sequence: DAEFRHDSGYEVHHQKLVFFAEDVGSNKGAIIGLMVGGVVIA
Specifications
Specifications
| For Use With (Application) | Electron Microscopy, Immunomicroscopy, In vitro Assay, In vivo Assay, Western Blot |
| Formulation | Dry powder. SeeReconstitution Instructions for re-suspension instructions/protocol. |
| Gene ID (Entrez) | 351 |
| Molecular Weight (g/mol) | M.W. Theoretical: 4.5 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -70C. Avoid freeze-thaw cycles. |
| Gene Alias | AAA, Abeta, ABPP, AD1, alpha-sAPP, Amyloid-beta precursor protein, APPI, Beta-Amyloid Peptide(1-42), CTFgamma, CVAP, PN2, PreA4 |
| Gene Symbol | APP |
| Purity or Quality Grade | >85% |
| Protein | beta Amyloid 42 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?