missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ beta Amyloid (1-42) Chimeric Recombinant Human Monoclonal Antibody (21F12)
Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551771
This item is not returnable.
View return policy
Description
This antibody recognizes amino acids 33-42 of amyloid beta.
Beta amyloid is a 40-43 amino acid peptide cleaved from amyloid precursor protein by a protease, gamma-secretase. Beta Amyloid (1-40) (Amyloid beta 40 or Abeta 40) together with beta Amyloid (1-42) (Amyloid beta 42 or Abeta 42) are two major C-terminal variants of the amyloid beta protein, constituting the majority of amyloid beta proteins. Both the Amyloid beta 40 and Amyloid beta 42 alloforms are major components of the extracellular plaques found in Alzheimer's disease brain tissue.
Specifications
| beta Amyloid (1-42) Chimeric | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| P05067 | |
| APP | |
| The original antibody was generated by immunizing A/J mice with a synthetic peptide (NH2-CYS-NH-CH2-(CH2)5-CO-GLMVGGVVIA-COOH) as an immunogen. | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 21F12 | |
| Unconjugated | |
| APP | |
| A beta 1 42; A ß 1 42; A4; AAA; AB42; Abeta; Abeta40; Abeta42; ABPP; AD1; Adap; AG; AICD-50; AICD-57; AICD-59; AID(50); AID(57); AID(59); Alpha-CTF; Alpha-secretase C-terminal fragment; Alzheimer disease amyloid A4 protein homolog; alzheimer disease amyloid protein; Amyloid; amyloid A4; Amyloid b; Amyloid beta; amyloid beta (A4) precursor protein; Amyloid beta 1 42; amyloid beta A4 protein; amyloid beta precursor protein; Amyloid intracellular domain 50; Amyloid intracellular domain 57; Amyloid intracellular domain 59; Amyloid precursor protein; Amyloid ß; Amyloid-beta A4 protein; Amyloid-beta precursor protein; Amyloid-beta protein 40; Amyloid-beta protein 42; Amyloidogenic glycoprotein; APP; APP-C57; APP-C59; APP-C99; APPI; appican; Aß 1 42; B APP42; BAPP42; beta-amyloid peptide; beta-amyloid peptide(1-40); beta-amyloid peptide(1-42); beta-amyloid precursor protein; betaApp; Beta-APP40; BetaAPP42; Beta-APP42; Beta-CTF; Beta-secretase C-terminal fragment; C31; C80; C83; C99; Cerebral vascular amyloid peptide; CTFgamma; Cvap; E030013M08Rik; Gamma-CTF(50); Gamma-CTF(57); Gamma-CTF(59); Gamma-secretase C-terminal fragment 50; Gamma-secretase C-terminal fragment 57; Gamma-secretase C-terminal fragment 59; N-APP; P3(40); P3(42); peptidase nexin-II; PN2; PN-II; PreA4; protease nexin II; protease nexin-II; S-APP-alpha; S-APP-beta; Soluble APP-alpha; Soluble APP-beta; testicular tissue protein Li 2; ß APP42; ßAPP42 | |
| Human | |
| Protein A | |
| RUO | |
| 351 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction