missing translation for 'onlineSavingsMsg'
Learn More

beta-Alanine, TRC

Product Code. 30329703
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30329703 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30329703 Supplier Toronto Research Chemicals Supplier No. TRCA6373201G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Beta-Alanine
CAS 107-95-9
Purity Grade Notes HPLC
Molecular Formula C3 H7 N O2
Quantity 1 g
Synonym Beta-Alanine, β-Alanine, 2-Carboxyethylamine, 3-Aminopropanoic acid, 3-Aminopropionic acid, 3-Azaniumylpropanoate, Abufene, NSC 7603, Propanoic acid, 3-amino-, β-Aminopropanoic acid, β-Aminopropionic acid, ω-Aminopropionic acid
InChI Formula InChI=1S/C3H7NO2/c4-2-1-3(5)6/h1-2,4H2,(H,5,6)
SMILES NCCC(=O)O
IUPAC Name 3-aminopropanoic acid
Molecular Weight (g/mol) 89.09
Formula Weight 89.0477
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.