missing translation for 'onlineSavingsMsg'
Learn More

beta-Alanine tert-butyl ester hydrochloride, 95%

Product Code. 15471509
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15471509 100 g 100g
15451509 5 g 5g
15461509 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15471509 Supplier Thermo Scientific Alfa Aesar Supplier No. H64346.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 58620-93-2 | C7H16ClNO2 | 181.66 g/mol

Useful intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful intermediate for pharmaceutical and organic synthesis.

Solubility
Clear solution (0.3g in 2ml MeOH).

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58620-93-2
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00058258
InChI Key DOMTZTVJNZKUNX-UHFFFAOYSA-N
Synonym tert-butyl 3-aminopropanoate hydrochloride, h-beta-ala-otbu.hcl, beta-alanine tert-butyl ester hydrochloride, h-beta-ala-otbu hcl, beta-alanine t-butyl ester hydrochloride, 3-amino-propionic acid tert-butyl ester hydrochloride, h-?-ala-otbu.hcl, h-, a-ala-otbu.hcl, h-beta-ala-otbu hydrochloride, tert-butyl 3-aminopropanoate-hcl
PubChem CID 16218890
IUPAC Name tert-butyl 3-aminopropanoate;hydrochloride
SMILES Cl.CC(C)(C)OC(=O)CCN

Specifications

Melting Point 167°C to 169°C
Linear Formula H2NCH2CH2COOC(CH3)3·HCl
Beilstein 6660122
Solubility Information Clear solution (0.3g in 2ml MeOH).
Quantity 100 g
Formula Weight 181.66
Percent Purity 95%
Physical Form Solid
Chemical Name or Material beta-Alanine tert-butyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.