missing translation for 'onlineSavingsMsg'
Learn More

Beta-Alanine Methyl Ester Hydrochloride, TRC

Product Code. 30316810
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30316810 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30316810 Supplier Toronto Research Chemicals Supplier No. TRCA5105602.5G
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material beta-Alanine Methyl Ester Hydrochloride
CAS 3196-73-4
Molecular Formula C4H10ClNO2
Quantity 2.5 g
Synonym (3-Methoxy-3-oxopropyl)ammonium Chloride, 3-Aminopropanoic Acid Methyl Ester Hydrochloride, 3-Aminopropionic Acid Methyl Ester Hydrochloride, 3-Aminopropionic Acid Methyl Ester Monohydrochloride, Methyl 3-Aminopropanoate Hydrochloride, Methyl 3-Aminopropionate Hydrochloride, Methyl beta-Alaninate Hydrochloride
InChI Formula InChI=1S/C4H9NO2.ClH/c1-7-4(6)2-3-5;/h2-3,5H2,1H3;1H
SMILES Cl.COC(=O)CCN
IUPAC Name methyl 3-aminopropanoate;hydrochloride
Molecular Weight (g/mol) 139.58
Formula Weight 139.04
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.