missing translation for 'onlineSavingsMsg'
Learn More

beta-Alanine ethyl ester hydrochloride, 98%

Product Code. 11403507
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403507 10 g 10g
11413507 50 g 50g
11423507 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11403507 Supplier Thermo Scientific Alfa Aesar Supplier No. A17277.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4244-84-2 | C5H12ClNO2 | 153.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4244-84-2
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.61
MDL Number MFCD00012909
InChI Key RJCGNNHKSNIUAT-UHFFFAOYSA-N
Synonym ethyl 3-aminopropanoate hydrochloride, beta-alanine ethyl ester hydrochloride, ethyl beta-alaninate hydrochloride, h-beta-ala-oet.hcl, h-b-ala-oet hcl, h-beta-ala-oet hcl, h-?-ala-oet.hcl, b-alanine ethyl ester hydrochloride, ethyl 3-aminopropionate hydrochloride, beta-alanine ethyl ester hcl
PubChem CID 458475
IUPAC Name ethyl 3-aminopropanoate;hydrochloride
SMILES Cl.CCOC(=O)CCN

Specifications

Melting Point 67°C to 72°C
Beilstein 3559095
Quantity 10 g
Formula Weight 153.61
Sensitivity Hygroscopic
Percent Purity 98%
Chemical Name or Material beta-Alanine ethyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.