missing translation for 'onlineSavingsMsg'
Learn More

beta-Alanine benzyl ester p-toluenesulfonate, 98%

Product Code. 11472938
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472938 10 g 10g
11482938 50 g 50g
11492938 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11472938 Supplier Thermo Scientific Alfa Aesar Supplier No. A18650.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 27019-47-2 | C10H14NO2 | 180.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27019-47-2
Molecular Formula C10H14NO2
Molecular Weight (g/mol) 180.23
MDL Number MFCD00039061
InChI Key CANCPUBPPUIWPX-UHFFFAOYSA-O
Synonym beta-alanine benzyl ester p-toluenesulfonate, beta-alanine benzyl ester p-toluenesulfonate salt, h-beta-ala-obzl.tosoh, h-beta-ala-obzl tos, benzyl 3-aminopropanoate 4-methylbenzenesulfonate, betaala-obzl.tosoh, pubchem12070, acmc-209gum, h-, a-ala-obzl.tosoh, h-ala-obzl p-tosylate
PubChem CID 13502100
SMILES [NH3+]CCC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 138°C to 139°C
IUPAC Name benzyl 3-aminopropanoate;4-methylbenzenesulfonic acid
Quantity 10 g
Formula Weight 351.42
Percent Purity 98%
Chemical Name or Material beta-Alanine benzyl ester p-toluenesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.