missing translation for 'onlineSavingsMsg'
Learn More

beta-Alaninamide hydrochloride, 96%, Thermo Scientific Chemicals

Catalog Number 15492529
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
64017-81-8
C3H9ClN2O
124.568
MFCD00060747
GAGJMOQGABUOBK-UHFFFAOYSA-N
22222288
3-aminopropanamide;hydrochloride
C(CN)C(=O)N.Cl
This item is not returnable. View return policy
64017-81-8
C3H9ClN2O
124.568
MFCD00060747
GAGJMOQGABUOBK-UHFFFAOYSA-N
22222288
3-aminopropanamide;hydrochloride
C(CN)C(=O)N.Cl

beta-Alaninamide hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
beta-Alaninamide hydrochloride is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol.

Notes
Air Sensitive. Store away from air. Protect from heat. Store under dry inert gas. Incompatible with heat , air and oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 149°C to 154°C
Beilstein 3909917
Synonym 3-aminopropanamide hydrochloride, 3-aminopropionamide hydrochloride, 3-aminopropanamide hcl, beta-alaninamide hydrochloride, h-beta-ala-nh2.hcl, h-beta-ala-nh2 hcl, 3-amino-propionamide hydrochloride, h-ala-nh2hcl, propanamide, 3-amino-, monohydrochloride, h-beta-ala-nh2hcl
Solubility Information Soluble in methanol.
Formula Weight 124.57
Percent Purity 96%
Quantity 5 g
Sensitivity Air sensitive
Physical Form Solid
Chemical Name or Material beta-Alaninamide hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'