missing translation for 'onlineSavingsMsg'
Learn More

Beta Actin Rabbit anti-Human, Mouse, Pig, Rat, Clone: 4H1, Proteintech

Rabbit Recombinant Monoclonal Antibody

Brand:  Proteintech 81115-1-RR-100UL

Product Code. 17711229

  • £189.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Beta Actin Recombinant Monoclonal antibody specifically detects Beta Actin in Human, Mouse, Pig, Rat samples. It is validated for IF, WB, ELISA
Specifications

Specifications

Beta Actin
Recombinant Monoclonal
1000 μg/mL
PBS with 0.02% sodium azide and 50% glycerol pH 7.3.
Rabbit
100 μL
60
Store at -20°C
Liquid
Immunofluorescence, Western Blot, ELISA
4H1
Unconjugated
BC013596
Protein A purification
Primary
Human, Mouse, Pig, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.