missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Actin Mouse anti-Human, Mouse, Plant, Porcine, Rat, Zebrafish, Clone: 7D2C10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60008-1-IG-150UL
This item is not returnable.
View return policy
Description
This antibody can recognize all the actins.
The product encoded by this gene belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actins being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. This actin is an alpha actin that is found in skeletal muscle. Mutations in this gene cause nemaline myopathy type 3, congenital myopathy with excess of thin myofilaments, congenital myopathy with cores, and congenital myopathy with fiber-type disproportion, diseases that lead to muscle fiber defects.Specifications
| beta Actin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P60709, P60710, P60711, P62736, P62737, P62738, P63259, P63260, P63261, P63267, P63268, P63269, P68032, P68033, P68035, P68133, P68134, P68136, P68137, Q6QAQ1 | |
| ACTA1, acta1b, acta2, ACTB, actb1, ACTC1, actc1b, Actg1, ACTG2 | |
| beta Actin Fusion Protein Ag0297 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Plant, Pig, Rat, Zebrafish | |
| Antibody | |
| IgM |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 7D2C10 | |
| Unconjugated | |
| ACTA1 | |
| ACTB, Actin, actin, beta, Actin, cytoplasmic 1, B actin, Beta actin, F actin | |
| Mouse | |
| Caprylic Acid Precipitation | |
| RUO | |
| 100152267, 100154254, 100520667, 11459, 11461, 11464, 11465, 11468, 11475, 25365, 287876, 29275, 29437, 322509, 397653, 407658, 414396, 57934, 58, 58114, 59, 60, 70, 71, 72, 733615, 81633, 81822 | |
| -20°C | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur