missing translation for 'onlineSavingsMsg'
Learn More

beta-Actin Antibody, HRP, Novus Biologicals™

Product Code. 18781144 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mL
Unit Size:
0.05mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18781144 0.05 mL 0.05mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18781144 Supplier Novus Biologicals Supplier No. NB600503H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 2 publications

beta-Actin Polyclonal specifically detects beta-Actin in Human, Mouse, Rat, Porcine, Avian, Bovine, Chinese Hamster, Fish, Primate, Rabbit samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta-Actin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS with No Preservative
Gene Accession No. P60709
Gene Alias ACTB, actin, beta, actin, cytoplasmic 1, B-Actin, Beta Actin, beta cytoskeletal actin, Beta-actin, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1
Gene Symbols ACTB
Host Species Rabbit
Immunogen This beta-Actin Antibody was made from a synthetic peptide made to an N-terminal region of human Beta Actin. [UniProt P60709]
Molecular Weight of Antigen 42 kDa
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Loading Controls, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 60
Test Specificity Human, Rat, Monkey, Mouse and Avian. Rabbit reactivity reported in the scientific literature (PMID: 23813946). Expected to cross-react with a wide range of species due to sequence homology.
Target Species Human, Mouse, Rat, Pig, Avian, Bovine, Chinese Hamster, Fish, Primate, Rabbit
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.