missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Actin Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-503F
This item is not returnable.
View return policy
Description
beta-Actin Polyclonal specifically detects beta-Actin in Human, Mouse, Rat, Porcine, Avian, Bovine, Chinese Hamster, Fish, Primate, Rabbit samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| beta-Actin | |
| Polyclonal | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| P60709 | |
| ACTB | |
| This beta-Actin Antibody was made from a synthetic peptide made to an N-terminal region of human Beta Actin. [UniProt P60709] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Rat, Monkey, Mouse and Avian. Rabbit reactivity reported in the scientific literature (PMID: 23813946). Expected to cross-react with a wide range of species due to sequence homology. | |
| Store at 4C in the dark. |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| ACTB, actin, beta, actin, cytoplasmic 1, B-Actin, Beta Actin, beta cytoskeletal actin, Beta-actin, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1 | |
| Rabbit | |
| 42 kDa | |
| 0.05 mL | |
| Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Loading Controls, Signal Transduction, Stem Cell Markers | |
| 60 | |
| Human, Mouse, Rat, Pig, Avian, Bovine, Chinese Hamster, Fish, Primate, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction