missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Actin Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-532R
This item is not returnable.
View return policy
Description
beta-Actin Polyclonal specifically detects beta-Actin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| beta-Actin | |
| Polyclonal | |
| Western Blot 1:5000-1:15000, Immunohistochemistry | |
| P60709 | |
| ACTB | |
| This beta-Actin Antibody maps to a region corresponding to the N-terminus of human Beta Actin. [UniProt# P60709] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ACTB, actin, beta, actin, cytoplasmic 1, B-Actin, Beta Actin, beta cytoskeletal actin, Beta-actin, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1 | |
| Rabbit | |
| 42 kDa | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Loading Controls, Signal Transduction, Stem Cell Markers | |
| 60 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction