missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Actin Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-532AF594
This item is not returnable.
View return policy
Description
beta-Actin Polyclonal specifically detects beta-Actin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| beta-Actin | |
| Polyclonal | |
| Western Blot, Immunohistochemistry | |
| ACTB, actin, beta, actin, cytoplasmic 1, B-Actin, Beta Actin, beta cytoskeletal actin, Beta-actin, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ACTB | |
| This beta-Actin Antibody maps to a region corresponding to the N-terminus of human Beta Actin. [UniProt# P60709] | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Loading Controls, Signal Transduction, Stem Cell Markers | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction