missing translation for 'onlineSavingsMsg'
Learn More

beta-Actin Antibody (8H10D10), DyLight 755, Novus Biologicals™

Product Code. 18419025 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419025 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419025 Supplier Novus Biologicals Supplier No. NBP147423IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

beta-Actin Monoclonal antibody specifically detects beta-Actin in Human, Mouse, Rat, Chinese Hamster, Primate samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunoblot, Flow (Intracellular)
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta-Actin
Applications Western Blot, Simple Western, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 8H10D10
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias ACTB, actin, beta, actin, cytoplasmic 1, B-Actin, Beta Actin, beta cytoskeletal actin, Beta-actin, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1
Host Species Mouse
Immunogen This beta-Actin Antibody (8H10D10) was made to a synthetic peptide corresponding to amino-terminal residues of human Beta Actin, conjugated to KLH. [UniProt# P60709]
Purification Method Ammonium sulfate precipitation
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Loading Controls, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 60
Target Species Human, Mouse, Rat, Chinese Hamster, Primate
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.