Learn More
Invitrogen™ beta-5 Tubulin Recombinant Rabbit Monoclonal Antibody (SR25-04)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA532008
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Microtubules, the major cytoskeletal elements found in all eukaryotic cells, consist of Tublin, which is a dimer of two 55kDa subunits: alpha and Beta. Microtubules play key roles in chromosome segregation in mitosis, intracellular transport, ciliary and flagellar bending, and structural support of the cytoskeleton. This antibody does not cause the 10-nm filaments to collapse into large lateral aggregates collecting in the cell periphery or tight juxtanuclear caps. It does not block microtubule assembly. Ab-3 does not inhibit polymerization or depolymerization of platelet tubulin in vitro. It blocks (by 70-80%) the ability of tubulin dimers (with GppNHp bound) to promote a stable inhibition of adenylyl cyclase.
Specifications
| beta-5 Tubulin | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
| P07437, P69897, P99024 | |
| TUBB, Tubb5 | |
| Synthetic peptide within Human beta Tubulin aa 308-357. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| SR25-04 | |
| Unconjugated | |
| TUBB | |
| AA408537; AI596182; B130022C14Rik; beta Ib tubulin; CDCBM6; CSCSC1; M(beta)5; M40; OK/SW-cl.56; tbb5; TUBB; TUBB1; TUBB5; tubulin beta; Tubulin beta chain; tubulin beta class I; tubulin beta-1 chain; tubulin beta-5 chain; tubulin, beta 5 class I; tubulin, beta class I; tubulin, beta polypeptide | |
| Rabbit | |
| Protein A | |
| RUO | |
| 203068, 22154, 29214 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.