Learn More
beta-3 Tubulin Monoclonal Antibody (2G10-TB3), eFluor™ 660, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 50-4510-80
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: The 2G10 monoclonal antibody recognizes beta tubulin class III, a member of the large family of tubulins that are components of the microtubules of the cytoskeletal network. Beta tubulin class III expression is exclusively found in neuronal cells of adult mammals. Although expression has been mainly found in neuronal cells, during fetal development, transient expression has been documented in both glial and neuronal precursors localized to the subventricular zone. Identification of beta tubulin class III expression is useful as a structural marker for developing and mature neural cells. Given the high degree of conservation of this protein across species, the 2G10 clone is expected to recognize human, mouse, rat, and bovine beta tubulin class III. Applications Reported: This 2G10-TB3 antibody has been reported for use in immunohistochemical staining of frozen and formalin-fixed paraffin embedded tissue sections, and immunocytochemistry. Applications Tested: This 2G10-TB3 antibody has been tested by immunocytochemistry of fixed and permeabilized cells and can be used at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor™ 660 is a replacement for Alexa Fluor™ 647. eFluor™ 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.
The betaIII-tubulin isoform is present dominantly in cells of neuronal origin and it is one of the earliest markers of neuronal differentiation. It is regarded as a specific probe for the cells of neuronal origin as well as for the tumours originating from these cells. The betaIII-tubulin is most abundant in cells of neuronal origin, but was also detected in Sertoli cells of the testis and transiently in non-neuronal embryonic tissues.Specifications
beta-3 Tubulin | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q13509, Q2T9S0, Q4QRB4, Q9ERD7 | |
TUBB3 | |
Affinity chromatography | |
RUO | |
10381, 22152, 246118, 768070 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
2G10-TB3 | |
eFluor 660 | |
TUBB3 | |
3200002H15Rik; beta 3 tubulin; beta tubulin 3; beta-4; betaIII-tubulin; CDCBM; CDCBM1; CFEOM3; CFEOM3A; class III beta-tubulin; FEOM3; I79_000194; LOC100341581; M(beta)3; M(beta)6; mc1R; melanocortin receptor 1; Neuron specific beta III Tubulin; neuron-specific class III beta-tubulin; TUBB; Tubb3; TUBB4; tubulin beta; tubulin beta 3 class III; Tubulin beta-3; tubulin beta-3 chain; Tubulin beta-4; Tubulin beta-4 chain; tubulin beta-III; tubulin, beta 3 class III; tubulin, beta, 3; tubulin, beta, 4; Unknown (protein for MGC:134115) | |
Mouse | |
25 μg | |
Primary | |
Bovine, Human, Mouse, Rat | |
Antibody | |
IgG2a κ |
For Research Use Only.