missing translation for 'onlineSavingsMsg'
Learn More

beta,3-Dinitrostyrene, 98%

Product Code. 11398725
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398725 1 g 1g
11308735 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398725 Supplier Thermo Scientific Alfa Aesar Supplier No. L12304.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 882-26-8 | C8H6N2O4 | 194.146 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 882-26-8
Molecular Formula C8H6N2O4
Molecular Weight (g/mol) 194.146
MDL Number MFCD00052063
InChI Key YOEGXQQUPVDQEE-SNAWJCMRSA-N
Synonym beta,3-dinitrostyrene, 1-nitro-3-2-nitrovinyl benzene, m,.beta.-dinitrostyrene, trans-m,beta-dinitrostyrene, m-2-nitrovinyl nitrobenzene, 3-nitro-b-nitrostyrene, styrene, m,beta-dinitro-, e, benzene, 1-nitro-3-2-nitroethenyl, 1-nitro-2-m-nitrophenyl ethylene, 1-nitro-3-e-2-nitroethenyl benzene
PubChem CID 5355017
IUPAC Name 1-nitro-3-[(E)-2-nitroethenyl]benzene
SMILES C1=CC(=CC(=C1)[N+](=O)[O-])C=C[N+](=O)[O-]

Specifications

Melting Point 125°C to 127°C
Quantity 1 g
Beilstein 2051290
Formula Weight 194.15
Percent Purity 98%
Chemical Name or Material beta,3-Dinitrostyrene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.