missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ beta-3 Defensin Polyclonal Antibody, Invitrogen™
GREENER_CHOICE

Rabbit Polyclonal Antibody

£155.00 - £386.00

Specifications

Antigen beta-3 Defensin
Concentration 0.5 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17713604
View Documents
Invitrogen™
PA5122203
20 μL
£155.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17703604
GREENER_CHOICE
View Documents
Invitrogen™
PA5122202
100 μL
£386.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 103, which has broad spectrum antimicrobial activity and may play an important role in innate epithelial defense. [provided by RefSeq, Oct 2014].
Specifications

Specifications

beta-3 Defensin
Western Blot
Unconjugated
Rabbit
Human, Pig
P81534
414325
Gly23-Lys67 (Accession P81534), with N-terminal His Tag.
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
DEFB103A
0.5 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
BD3; BD-3; beta defensin 3; beta defensin-3; beta-defensin 103; Beta-defensin 3; DEFB103; DEFB103A; DEFB103B; DEFB3; DEFB-3; defensin beta 103A; defensin, beta 103; defensin, beta 103A; defensin, beta 3; defensin-like protein; HBD3; hBD-3; HBP3; HBP-3
DEFB103A
IgG
Antigen affinity chromatography, Protein A
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.