Learn More
beta-3 Defensin Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 500-P241BT-1MG
Description
AA Sequence of recombinant protein: GIINTLQKYY CRVRGGRCAV LSCLPKEEQI GKCSTRGRKC CRRKK. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human BD-3. Anti-Human BD-3-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect hBD-3 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human BD-3 (500-P241) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant hBD-3. Western Blot: To detect hBD-3 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hBD-3 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P241BT-1MG will be provided as 2 x 500 μg
Defensins form a family of microbicidal and cytotoxic peptides made by neutrophils. Members of the defensin family are highly similar in protein sequence. This gene encodes defensin, beta 103, which has broad spectrum antimicrobial activity and may play an important role in innate epithelial defense. [provided by RefSeq, Oct 2014].
Specifications
| beta-3 Defensin | |
| Polyclonal | |
| Biotin | |
| DEFB103A | |
| BD3; BD-3; beta defensin 3; beta defensin-3; beta-defensin 103; Beta-defensin 3; DEFB103; DEFB103A; DEFB103B; DEFB3; DEFB-3; defensin beta 103A; defensin, beta 103; defensin, beta 103A; defensin, beta 3; defensin-like protein; HBD3; hBD-3; HBP3; HBP-3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 414325 | |
| -20°C |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P81534 | |
| DEFB103A | |
| E.coli-derived Recombinant Human BD-3 | |
| 2 x 500 μg | |
| Primary | |
| Human | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.