missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ beta-3 Adrenergic Receptor Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA5142837
This item is not returnable.
View return policy
Description
Additional Information: The immunizing peptide represents part of an intracellular domain. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
beta 3 Adrenergic Receptor is located mainly in adipose tissue and is involved in the regulation of lipolysis and thermogenesis. Beta adrenergic receptors are involved in the epinephrine and norepinephrine-induced activation of adenylate cyclase through the action of G proteins. beta 3 Adrenergic Receptor expression has been documented in adipose tissues, heart, and in smooth muscle of digestive and urinary tract organs (bladder, colon, small intestine, stomach, ureter). Utilization of alternate promoters and/or 3-prime untranslated regions may result in tissue-specific regulation of the expression of this protein.
Specifications
| beta-3 Adrenergic Receptor | |
| Polyclonal | |
| Unconjugated | |
| ADRB3 | |
| ADRB; ADRB3; Adrb-3; Adrb3r; adrenergic receptor, beta 3; adrenergic, beta-3-, receptor; adrenoceptor beta 3; B3AR; B3bar; beta 3 adrenoceptor; beta 3-AR; beta-3 adrenergic receptor; Beta-3 adrenoceptor; beta-3 adrenoreceptor; Beta-3 AR; beta3-adrenergic receptor; beta3-Adrenoreceptor; BETA3AR; Beta3-AR; FLJ99960 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 155 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P13945 | |
| ADRB3 | |
| Peptide with sequence C-EARQSPPLNRFD (aa 379-390). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction