missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-3 Adrenergic Receptor Polyclonal Antibody

Description
beta-3 Adrenergic Receptor Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | beta-3 Adrenergic Receptor |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | ADRB3 |
| Gene Accession No. | P13945 |
| Gene Alias | ADRB; ADRB3; Adrb-3; Adrb3r; adrenergic receptor, beta 3; adrenergic, beta-3-, receptor; adrenoceptor beta 3; B3AR; B3bar; beta 3 adrenoceptor; beta 3-AR; beta-3 adrenergic receptor; Beta-3 adrenoceptor; beta-3 adrenoreceptor; Beta-3 AR; beta3-adrenergic receptor; beta3-Adrenoreceptor; BETA3AR; Beta3-AR; FLJ99960 |
| Gene Symbols | ADRB3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?