missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ beta-3 Adrenergic Receptor Monoclonal Antibody (493324)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA562027
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
beta 3 Adrenergic Receptor is located mainly in adipose tissue and is involved in the regulation of lipolysis and thermogenesis. Beta adrenergic receptors are involved in the epinephrine and norepinephrine-induced activation of adenylate cyclase through the action of G proteins. beta 3 Adrenergic Receptor expression has been documented in adipose tissues, heart, and in smooth muscle of digestive and urinary tract organs (bladder, colon, small intestine, stomach, ureter). Utilization of alternate promoters and/or 3-prime untranslated regions may result in tissue-specific regulation of the expression of this protein.
Specifications
| beta-3 Adrenergic Receptor | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and no preservative | |
| P13945 | |
| ADRB3 | |
| Human embryonic kidney cell line HEK293-derived transfected with human beta-3 Adrenergic R/ADRB3, Met1-Ser408. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunocytochemistry | |
| 493324 | |
| Unconjugated | |
| ADRB3 | |
| ADRB; ADRB3; Adrb-3; Adrb3r; adrenergic receptor, beta 3; adrenergic, beta-3-, receptor; adrenoceptor beta 3; B3AR; B3bar; beta 3 adrenoceptor; beta 3-AR; beta-3 adrenergic receptor; Beta-3 adrenoceptor; beta-3 adrenoreceptor; Beta-3 AR; beta3-adrenergic receptor; beta3-Adrenoreceptor; BETA3AR; Beta3-AR; FLJ99960 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 155 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction