missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-3 Adrenergic Receptor (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577284
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20C upon receipt. Reconstitute using 50 uL deionized water.
beta 3 Adrenergic Receptor is located mainly in adipose tissue and is involved in the regulation of lipolysis and thermogenesis. Beta adrenergic receptors are involved in the epenephrine and norepinephrine-induced activation of adenylate cyclase through the action of G proteins. beta 3 Adrenergic Receptor expression has been documented in adipose tissues, heart, and in smooth muscle of digestive and urinary tract organs (bladder, colon, small intestine, stomach, ureter). Utilization of alternate promoters and/or 3-prime untranslated regions may result in tissue-specific regulation of the expression of this protein.
Specifications
| beta-3 Adrenergic Receptor (extracellular) | |
| Polyclonal | |
| Unconjugated | |
| Adrb3 | |
| beta3-Adrenoreceptor, ADRB3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11556, 25645 | |
| -20°C | |
| Lyophilized |
| Immunofluorescence, Immunohistochemistry, Western Blot | |
| 0.8 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| P25962, P26255 | |
| Adrb3 | |
| Peptide DAEAQESH*SNPRC, corresponding to amino acid residues 180-192 of mouse beta3 -Adrenoceptor with replacement of cysteine 186 (C186 ) with serine (*S) | |
| 50 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction