missing translation for 'onlineSavingsMsg'
Learn More

beta-2 Microglobulin Recombinant Rabbit Monoclonal Antibody (SU35-05)
GREENER_CHOICE

beta-2 Microglobulin Recombinant Rabbit Monoclonal Antibody (SU35-05)

Brand:  Invitrogen MA545029

Product Code. 17806222

  • £418.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

beta-2 Microglobulin Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IP

TRUSTED_SUSTAINABILITY
Specifications

Specifications

beta-2 Microglobulin
Recombinant Monoclonal
1 mg/mL
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
P01887, P07151, P61769
B2m
Synthetic peptide within C-terminal human beta 2 Microglobulin.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Western Blot, Immunocytochemistry, Western Blot
SU35-05
Unconjugated
B2m
B2m; B2-microglobulin; b2m-W01; b2m-W03; b2m-Z01; b2m-Z02; b2m-Z03; Beta 2 microglobin; Beta 2 microglobulin; beta 2-microglobulin; beta chain of MHC class I molecules; Beta2 microglobulin; beta-2 microglobulin; beta-2 microglobulin preprotein; beta2m; beta2-m; beta-2-microglobin; beta-2-microglobulin; beta-2-microglobulin C-terminal fragment (55 AA); Beta-2-microglobulin form pI 5.3; B-F; CDABP0092; HDCMA22P; IMD43; Lactollin; Ly-m11; MHC Class I; microglobulin beta-2; Orla-B2m
Rabbit
Protein A
RUO
12010, 24223, 567
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.