missing translation for 'onlineSavingsMsg'
Learn More

beta 2-Microglobulin, Rabbit anti-Human, Clone: BBM.1, Novus Biologicals™

Rabbit Monoclonal Antibody

£198.00 - £510.00

Specifications

Antigen beta 2-Microglobulin
Clone BBM.1
Concentration 1 mg/mL
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Applications Western Blot, Immunohistochemistry, Immunofluorescence
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18378910
View Documents
Novus Biologicals
NBP2-81045-0.025MG
0.025 mg
£198.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18377100
View Documents
Novus Biologicals
NBP2-81045-0.2MG
0.2 mg
£510.00
0.20mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

beta 2-Microglobulin Monoclonal specifically detects beta 2-Microglobulin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

beta 2-Microglobulin
1 mg/mL
Western Blot, Immunohistochemistry, Immunofluorescence
Unconjugated
Rabbit
Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology
PBS
567
Molt 4, a human T cell line.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
BBM.1
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Monoclonal
Purified
RUO
Human
beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin
B2M
IgG κ
Protein A purified
Binds human beta 2-Microglobulin, a component of MHC class I molecules, which are present on all nucleated cells (excludes red blood cells) and involved in the presentation of peptide antigens to the immune system.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.