Learn More
beta 2-Microglobulin Antibody (B2M/961), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-44471-0.1mg
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
beta 2-Microglobulin Monoclonal specifically detects beta 2-Microglobulin in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Cell Surface).
Specifications
| beta 2-Microglobulin | |
| Monoclonal | |
| B2M | |
| 12 kDa | |
| Store at 4C. |
| Flow Cytometry | |
| B2M/961 | |
| Full length recombinant human B2M protein | |
| 0.1 mg | |
| IgG2b κ |