missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta 2-Microglobulin Antibody (B2M/961), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-44471-0.02MG
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
beta 2-Microglobulin Monoclonal specifically detects beta 2-Microglobulin in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Cell Surface).
Specifications
| beta 2-Microglobulin | |
| Monoclonal | |
| 0.2mg/mL | |
| Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml | |
| P61769 | |
| B2M | |
| Full length recombinant human B2M protein | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Recognizes a protein of 12kDa, identified as beta-2 microglobulin. Major histocompatibility complex (MHC) class 1 molecules bind to antigens for presentation on the surface of cells. The proteasome is responsible for producing these antigens from the components of foreign pathogens. MHC class 1 molecules consist of an alpha heavy chain that contains three subdomains (alpha1, alpha2, alpha3) and a non-covalent associating light chain, known as beta-2-Microglobulin. beta-2-Microglobulin associates with the alpha3 subdomain of the alpha heavy chain and forms an immunoglobulin domain-like structure that mediates proper folding and expression of MHC class 1 molecules. The alpha1 and alpha2 domains of the alpha heavy chain form the peptide antigen-binding cleft. Mutations in the beta-2-Microglobulin gene can enhance the progression of malignant melanoma phenotypes. | |
| Store at 4C. | |
| IgG2b κ |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| B2M/961 | |
| Unconjugated | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin | |
| Mouse | |
| 12 kDa | |
| 0.02 mg | |
| Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology | |
| 567 | |
| Human, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction